(2-Chloro-4-methoxyphenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Chloro-4-methoxyphenyl)acetonitrile

021049-500MG 021049-2.5G
101811-272EA 103.08 USD
101811-272 101811-274
(2-Chloro-4-methoxyphenyl)acetonitrile ≥95%
(2-Chloro-4-methoxyphenyl)acetonitrile

Hazard: Irritant

Formula: C₉H₈ClNO
MW: 181.62 g/mol
Melting Pt: 42…44 °C
MDL Number: MFCD08445710
CAS Number: 170737-93-6

Order Now

Specification Test Results

Melting point 42-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR