2,5-Dimethoxybenzenesulfonamide ≥97%

Supplier: Matrix Scientific

020956-1G 020956-5G
101811-090EA 44.62 USD
101811-090 101811-092
2,5-Dimethoxybenzenesulfonamide ≥97%
2,5-Dimethoxybenzenesulphonamide

Hazard: Irritant

Formula: C₈H₁₁NO₄S
Melting Pt: 150-152°C
MDL Number: MFCD00833415
CAS Number: 19116-90-6

Order Now

Specification Test Results

Melting point 150-152°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR