4-Methyl-3-nitrobenzenesulfonamide ≥97%
Supplier: Matrix Scientific
020955-1G
020955-5G
101811-086EA
90.77
USD
101811-086
101811-088
4-Methyl-3-nitrobenzenesulfonamide ≥97%
4-Methyl-3-nitrobenzenesulfonamide
Hazard: Irritant
Formula:
C₇H₈N₂O₄S MW: 216.22 g/mol Melting Pt: 144 °C |
MDL Number:
MFCD00460611 CAS Number: 6949-23-1 |
Specification Test Results
Melting point | 144°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...