1,2,4-Triazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

011382-1G 011382-5G
101800-538EA 15.38 USD
101800-538 101800-540
1,2,4-Triazole-3-carboxylic acid ≥95%
1,2,4-Triazole-3-carboxylic acid

Hazard: Irritant

Formula: C₃H₃N₃O₂
Melting Pt: 132…136 °C
MDL Number: MFCD00078268
CAS Number: 4928-87-4

Order Now

Specification Test Results

Melting point 132-136°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR