3-Cyano-5-fluorobenzoic acid ≥98%
Supplier: Matrix Scientific
011291-5G
011291-25G
101800-364EA
67.69
USD
101800-364
101800-366
3-Cyano-5-fluorobenzoic acid ≥98%
3-Cyano-5-fluorobenzoic acid
Hazard: Irritant
Formula:
C₈H₄FNO₂ Melting Pt: 172…173 °C |
MDL Number:
MFCD05663717 CAS Number: 327056-74-6 |
Specification Test Results
Melting point | 172-173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...