(2-Bromo-5-fluorophenyl)acetic acid ≥98%
Supplier: Matrix Scientific
011281-1G
011281-5G
101800-320EA
20
USD
101800-320
101800-322
(2-Bromo-5-fluorophenyl)acetic acid ≥98%
(2-Bromo-5-fluorophenyl)acetic acid
Hazard: Irritant
Formula:
C₈H₆BrFO₂ Melting Pt: 113-114°C |
MDL Number:
MFCD03094440 CAS Number: 739336-26-6 |
Specification Test Results
Melting point | 113-114°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...