5-Bromo-2-fluorophenylacetic acid ≥96%
Supplier: Matrix Scientific
011280-5G
011280-25G
101800-316EA
23.08
USD
101800-316
101800-318
5-Bromo-2-fluorophenylacetic acid ≥96%
5-Bromo-2-fluorophenylacetic acid
Hazard: Irritant
Formula:
C₈H₆BrFO₂ MW: 233.0345 g/mol Melting Pt: 111-112°C |
MDL Number:
MFCD06246020 CAS Number: 883514-21-4 |
Specification Test Results
Melting point | 111-112°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...