Azetidin-3-ol hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxyazetidine hydrochloride

011272-5G 011272-25G 011272-100G
101800-290EA 15.38 USD
101800-290 101800-292 101800-294
Azetidin-3-ol hydrochloride ≥97%
Azetidin-3-ol hydrochloride

Hazard: Irritant

Formula: C₃H₈ClNO
Melting Pt: 90…92 °C
MDL Number: MFCD02683887
CAS Number: 18621-18-6

Order Now

Specification Test Results

Melting point 90-92°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR