6-(Benzyloxy)-1H-indole ≥98%

Supplier: Matrix Scientific

011269-1G 011269-5G
101800-282EA 21.54 USD
101800-282 101800-284
6-(Benzyloxy)-1H-indole ≥98%
6-(Benzyloxy)-1H-indole

Hazard: Irritant/Store in Freezer

Formula: C15H13NO
Melting Pt: 114…116 °C
MDL Number: MFCD00053554
CAS Number: 15903-94-3

Order Now

Specification Test Results

Melting point 114-116°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR