2-Methoxyisonicotinic acid ≥98%

Supplier: Matrix Scientific

011266-5G 011266-25G
101800-274EA 29.23 USD
101800-274 101800-276
2-Methoxyisonicotinic acid ≥98%
2-Methoxyisonicotinic acid

Hazard: Irritant

Formula: C₇H₇NO₃
Melting Pt: 197-200°C
MDL Number: MFCD04115718
CAS Number: 105596-63-2

Order Now

Specification Test Results

Melting point 197-200°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR