2-Amino-6-methoxy-3-nitropyridine ≥98%

Supplier: Matrix Scientific

011260-5G 011260-25G
101800-256EA 15.38 USD
101800-256 101800-258
2-Amino-6-methoxy-3-nitropyridine ≥98%
2-Amino-6-methoxy-3-nitropyridine

Hazard: Irritant

Formula: C₆H₇N₃O₃
Melting Pt: 170…172 °C
MDL Number: MFCD03206481
CAS Number: 73896-36-3

Order Now

Specification Test Results

Melting point 170-172°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR