Methyl-2-bromoisonicotinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-bromoisonicotinate

011236-5G 011236-25G
101800-188EA 44.62 USD
101800-188 102832-834
Methyl-2-bromoisonicotinate ≥97%
Methyl 2-bromoisonicotinate

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.0320 g/mol
Melting Pt: 35-36°C
MDL Number: MFCD03791265
CAS Number: 26156-48-9

Order Now

Specification Test Results

Melting point 35-36°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR