3,5-Dichloro-4-pyridone ≥98%
Supplier: Matrix Scientific
Synonyms:
3,5-Dichloropyridin-4(1H)-one
011215-25G
011215-100G
101800-138EA
89.23
USD
101800-138
101800-140
3,5-Dichloro-4-pyridone ≥98%
3,5-Dichloro-4-pyridone
Hazard: Irritant
Formula:
C₅H₃Cl₂NO Melting Pt: 320-325°C |
MDL Number:
MFCD00051673 CAS Number: 17228-70-5 |
Specification Test Results
Melting point | 320-325°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...