4-(Methylthio)-m-cresol ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Methyl-4-(methylthio)phenol
011176-5G
011176-25G
011176-100G
101800-080EA
15.38
USD
101800-080
101800-082
101800-084
4-(Methylthio)-m-cresol ≥97%
4-(Methylthio)-m-cresol
TCA listed: T
Hazard: Irritant/Stench
Formula:
C₈H₁₀OS MW: 154.23 g/mol |
MDL Number:
MFCD00045773 CAS Number: 3120-74-9 |
Specification Test Results
Melting point | 49-53°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...