2-(Trifluoromethyl)thioxanthen-9-one ≥97%
Supplier: Matrix Scientific
011163-5G
011163-25G
101800-028EA
95.38
USD
101800-028
101800-030
2-(Trifluoromethyl)thioxanthen-9-one ≥97%
2-(Trifluoromethyl)thioxanthen-9-one
Hazard: Irritant
Formula:
C₁₄H₇F₃OS MW: 280.26 g/mol Melting Pt: 143…145 °C |
MDL Number:
MFCD00079785 CAS Number: 1693-28-3 |
Specification Test Results
Melting point | 143-145°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...