3,5-Dichloropyridine-2-carbonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
3,5-Dichloropicolinonitrile, 3,5-Dichloro-2-cyanopyridine
011120-5G
011120-25G
101799-920EA
26.15
USD
101799-920
101799-922
3,5-Dichloropyridine-2-carbonitrile ≥97%
3,5-Dichloropyridine-2-carbonitrile
Hazard: Irritant
Formula:
C₆H₂Cl₂N₂ Melting Pt: 101…103 °C |
MDL Number:
MFCD03788758 CAS Number: 85331-33-5 |
Specification Test Results
Melting point | 101-103°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...