4,8-Dichloro-2-(trifluoromethyl)quinoline ≥97%
Supplier: Matrix Scientific
004986-1G
004986-5G
101788-118EA
87.69
USD
101788-118
101788-120
4,8-Dichloro-2-(trifluoromethyl)quinoline ≥97%
4,8-Dichloro-2-(trifluoromethyl)quinoline
Hazard: Irritant
Formula:
C₁₀H₄Cl₂F₃N MW: 266.0467 g/mol Melting Pt: 60-63°C |
MDL Number:
MFCD00276590 CAS Number: 18706-35-9 |
Specification Test Results
Melting point | 60-63°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...