3-(3-Nitrophenyl)propionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Nitrohydrocinnamic acid

004938-1G 004938-5G
101788-056EA 13.85 USD
101788-056 101788-058
3-(3-Nitrophenyl)propionic acid ≥98%
3-(3-Nitrophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₉NO₄
MW: 195.1721 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD01310835
CAS Number: 1664-57-9

Order Now

Specification Test Results

Melting point 110-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR