1-(4-Nitrophenyl)imidazoline-2-thione
Supplier: Matrix Scientific
004935-1G
004935-5G
101788-042EA
50.77
USD
101788-042
101788-044
1-(4-Nitrophenyl)imidazoline-2-thione
1-(4-Nitrophenyl)imidazoline-2-thione
Hazard: Irritant
Formula:
C₉H₇N₃O₂S Melting Pt: 232…233 °C (dec.) |
MDL Number:
MFCD00060484 CAS Number: 6857-35-8 |
Specification Test Results
Melting point | 232-233°C(d |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...