1-(4-Bromophenyl)imidazoline-2-thione ≥98%
Supplier: Matrix Scientific
004933-1G
004933-5G
101788-034EA
78.46
USD
101788-034
101788-036
1-(4-Bromophenyl)imidazoline-2-thione ≥98%
1-(4-Bromophenyl)imidazoline-2-thione
Hazard: Irritant
Formula:
C₉H₇BrN₂S Melting Pt: 244…246 °C (dec.) |
MDL Number:
MFCD00060482 CAS Number: 17452-23-2 |
Specification Test Results
Melting point | 244-246°C(d |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...