4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile

Supplier: Matrix Scientific

Synonyms: 4-((5-(Trifluoromethyl)pyridin-2-yl)oxy)benzonitrile

004925-1G 004925-5G
101788-010EA 60 USD
101788-010 101788-012
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₇F₃N₂O
MW: 264.21 g/mol
Melting Pt: 83…85 °C
MDL Number: MFCD00173780
CAS Number: 175277-01-7

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR