9-Fluorenone-2,7-dicarboxylic acid, Technical Grade
Supplier: Matrix Scientific
Synonyms:
9-Oxo-9H-fluorene-2,7-dicarboxylic acid
004908-1G
004908-5G
101787-984EA
169.23
USD
101787-984
101787-986
9-Fluorenone-2,7-dicarboxylic acid, Technical Grade
9-Fluorenone-2,7-dicarboxylic acid
Hazard: Irritant
Formula:
C₁₅H₈O₅ MW: 268.23 g/mol Melting Pt: 407 °C |
MDL Number:
MFCD00016355 CAS Number: 792-26-7 |
Specification Test Results
Melting point | 407°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...