2-Mercapto-5-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethyl)-2(1H)-pyridinethione, 5-(Trifluoromethyl)-2-pyridinethiol, 5-(Trifluoromethyl)pyridine-2(1H)-thione

004803-5G 004803-25G
101787-838EA 26.15 USD
101787-838 101787-840
2-Mercapto-5-(trifluoromethyl)pyridine ≥95%
2-Mercapto-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄F₃NS
MW: 179.16 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00128893
CAS Number: 76041-72-0

Order Now

Specification Test Results

Melting point 159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR