N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine

Supplier: Matrix Scientific

Synonyms: 1-(2-Nitro-4-(trifluoromethyl)phenyl)piperidine

004794-5G 004794-25G
101787-826EA 43.08 USD
101787-826 101787-828
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₃F₃N₂O₂
Melting Pt: 48-50°C
MDL Number: MFCD00052908
CAS Number: 1692-79-1

Order Now

Specification Test Results

Melting point 48-50°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR