N-[2-Nitro-4-(trifluoromethyl)phenyl]morpholine

Supplier: Matrix Scientific

004792-1G 004792-5G
101787-818EA 60 USD
101787-818 101787-820
N-[2-Nitro-4-(trifluoromethyl)phenyl]morpholine
N-[2-Nitro-4-(trifluoromethyl)phenyl]morpholine

Hazard: Irritant

Formula: C₁₁H₁₁F₃N₂O₃
Melting Pt: 34-36°C
MDL Number: MFCD00052375
CAS Number: 62054-72-2

Order Now

Specification Test Results

Melting point 34-36°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR