Benzaldehyde azine

Supplier: Matrix Scientific

004723-5G 004723-25G
101787-744EA 21.54 USD
101787-744 101787-746
Benzaldehyde azine
Benzaldehyde azine

Hazard: Irritant/Irritant-Harmful

Formula: C₁₄H₁₂N₂
Melting Pt: 91…93 °C
MDL Number: MFCD00016876
CAS Number: 588-68-1

Order Now

Specification Test Results

Melting point 91-93°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR