2,4-Diamino-5-fluoroquinazoline

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2,4-diaminoquinazoline

004672-1G 004672-5G
101787-662EA 52.31 USD
101787-662 101787-664
2,4-Diamino-5-fluoroquinazoline
2,4-Diamino-5-fluoroquinazoline

Hazard: Irritant

Formula: C₈H₇FN₄
MW: 178.17 g/mol
Melting Pt: 245-248°C
MDL Number: MFCD00221785
CAS Number: 119584-70-2

Order Now

Specification Test Results

Melting point 245-248°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR