1,2-Diamino-4-chloro-5-fluorobenzene ≥97%

Supplier: Matrix Scientific

004670-1G 004670-5G 004670-25G
101787-652EA 24.62 USD
101787-652 101787-654 101787-656
1,2-Diamino-4-chloro-5-fluorobenzene ≥97%
1,2-Diamino-4-chloro-5-fluorobenzene

Hazard: Irritant

Formula: C₆H₆ClFN₂
Melting Pt: 110…112 °C
MDL Number: MFCD00042485
CAS Number: 139512-70-2

Order Now

Specification Test Results

Melting point 110-112°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR