2-Bromocinnamic acid ≥98%
Supplier: Matrix Scientific
004659-5G
004659-25G
004659-100G
101787-638EA
21.54
USD
101787-638
101787-640
102832-760
2-Bromocinnamic acid ≥98%
2-Bromocinnamic acid
Hazard: Irritant
Formula:
C₉H₇BrO₂ Melting Pt: 215…216 °C |
MDL Number:
MFCD00016836 CAS Number: 7499-56-1 |
Specification Test Results
Melting point | 215-216°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...