2,3-Dibromo-5-thiophenecarboxylic acid hydrazide

Supplier: Matrix Scientific

004559-1G 004559-5G
101787-482EA 63.08 USD
101787-482 101787-484
2,3-Dibromo-5-thiophenecarboxylic acid hydrazide
2,3-Dibromo-5-thiophenecarboxylic acid hydrazide

Hazard: Irritant

Formula: C₅H₄Br₂N₂OS
MW: 299.98 g/mol
Melting Pt: 201…203 °C
MDL Number: MFCD00456543
CAS Number: 171851-25-5

Order Now

Specification Test Results

Melting point 201-203°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR