2,4-Dibromo-5-methylanisole ≥98%

Supplier: Matrix Scientific

004558-5G 004558-25G
101787-478EA 70.77 USD
101787-478 101787-480
2,4-Dibromo-5-methylanisole ≥98%
2,4-Dibromo-5-methylanisole

Hazard: Irritant

Formula: C₈H₈Br₂O
Melting Pt: 74…76 °C
MDL Number: MFCD00017771
CAS Number: 5456-94-0

Order Now

Specification Test Results

Melting point 74-76°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR