Ethyl-2-amino-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

004517-1G 004517-5G
101787-374EA 20 USD
101787-374 101787-376
Ethyl-2-amino-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate ≥97%
Ethyl-2-amino-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

TCA listed: T
Hazard: Irritant

Formula: C₇H₇F₃N₂O₂S
Melting Pt: 170…172 °C
MDL Number: MFCD00153147
CAS Number: 344-72-9

Order Now

Specification Test Results

Melting point 170-172°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR