1-Phenyl-1H-imidazole-2-thiol ≥98%

Supplier: Matrix Scientific

004506-1G 004506-5G
101787-348EA 84.62 USD
101787-348 101787-350
1-Phenyl-1H-imidazole-2-thiol ≥98%
1-Phenyl-1H-imidazole-2-thiol

Hazard: Irritant

Formula: C₉H₈N₂S
Melting Pt: 184…187 °C
MDL Number: MFCD00127929
CAS Number: 17452-09-4

Order Now

Specification Test Results

Melting point 184-187°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR