8-(Chloromethyl)-6-fluorobenzo-1,3-dioxane

Supplier: Matrix Scientific

Synonyms: 8-(Chloromethyl)-6-fluoro-4H-1,3-benzodioxine

004480-250MG 004480-1G
101787-306EA 106.15 USD
101787-306 101787-308
8-(Chloromethyl)-6-fluorobenzo-1,3-dioxane
8-(Chloromethyl)-6-fluorobenzo-1,3-dioxane

Hazard: Irritant

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Melting Pt: 81…84 °C
MDL Number: MFCD00052603
CAS Number: 131728-94-4

Order Now

Specification Test Results

Melting point 81-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR