3-(2,5-Dimethoxyphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(2,5-Dimethoxyphenyl)propanoic acid

004464-1G 004464-5G
101787-276EA 29.23 USD
101787-276 101787-278
3-(2,5-Dimethoxyphenyl)propionic acid ≥98%
3-(2,5-Dimethoxyphenyl)propionic acid

TCA listed: T
Hazard: Irritant

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Melting Pt: 62-65°C
MDL Number: MFCD00060325
CAS Number: 10538-49-5

Order Now

Specification Test Results

Melting point 62-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR