1-(3-Trifluoromethylphenyl)imidazoline-2-thione ≥98%
Supplier: Matrix Scientific
Synonyms:
1,3-Dihydro-1-[3-(trifluoromethyl)phenyl]-2H-imidazole-2-thione
004408-1G
004408-5G
101787-190EA
38.46
USD
101787-190
101787-192
1-(3-Trifluoromethylphenyl)imidazoline-2-thione ≥98%
1-(3-Trifluoromethylphenyl)imidazoline-2-thione
Hazard: Irritant
Formula:
C₁₀H₇F₃N₂S MW: 244.24 g/mol Melting Pt: 157…159 °C |
MDL Number:
MFCD00041202 CAS Number: 17452-08-3 |
Specification Test Results
Melting point | 157-159°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...