3H-Heptafluoro-2,2,4,4-tetrahydroxypentane ≥97%

Supplier: Matrix Scientific

Synonyms: 1,1,1,3,5,5,5-Heptafluoroacetylacetone dihydrate

004373-1G 004373-5G
101787-120EA 176.92 USD
101787-120 101787-122
3H-Heptafluoro-2,2,4,4-tetrahydroxypentane ≥97%
3H-Heptafluoro-2,2,4,4-tetrahydroxypentane

Hazard: Irritant

Formula: C₅H₅F₇O₄
MW: 262.08 g/mol
Melting Pt: 113…114 °C
MDL Number: MFCD02093308
CAS Number: 77953-71-0

Order Now

Specification Test Results

Melting point 113-114°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR