3',5'-Difluoro-2'-hydroxyacetophenone ≥98%
Supplier: Matrix Scientific
004372-1G
004372-5G
101787-116EA
58.46
USD
101787-116
101787-118
3',5'-Difluoro-2'-hydroxyacetophenone ≥98%
3',5'-Difluoro-2'-hydroxyacetophenone
Hazard: Irritant
Formula:
C₈H₆F₂O₂ Melting Pt: 108…110 °C |
MDL Number:
MFCD00042476 CAS Number: 140675-42-9 |
Specification Test Results
Melting point | 108-110°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...