Pseudopelletierine ≥98%
Supplier: Matrix Scientific
Synonyms:
9-Methyl-9-azabicyclo[3.3.1]nonan-3-one
004331-1G
004331-5G
101787-088EA
75.38
USD
101787-088
101787-090
Pseudopelletierine ≥98%
Pseudopelletierine
Hazard: Irritant
Formula:
C₉H₁₅NO MW: 153.23 g/mol Melting Pt: 63…65 °C |
MDL Number:
MFCD00078161 CAS Number: 552-70-5 |
Specification Test Results
Melting point | 63-65°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...