3,4-Methylenedioxyphenethylamine hydrochloride ≥98%
Supplier: Matrix Scientific
004301-1G
004301-5G
101787-068EA
192.31
USD
101787-068
101787-070
3,4-Methylenedioxyphenethylamine hydrochloride ≥98%
3,4-Methylenedioxyphenethylamine hydrochloride
Hazard: Irritant/Irritant-Harmful/Moisture Sensitive
Formula:
C₉H₁₂ClNO₂ Melting Pt: 213…214 °C |
MDL Number:
MFCD00035193 CAS Number: 1653-64-1 |
Specification Test Results
Melting point | 213-214°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...