5-Bromo-2-thiophenesulfonamide

Supplier: Matrix Scientific

Synonyms: 5-Bromothiophene-2-sulphonamide

004203-5G 004203-25G
101786-946EA 20 USD
101786-946 101786-948
5-Bromo-2-thiophenesulfonamide
5-Bromo-2-thiophenesulfonamide

Hazard: Irritant

Formula: C₄H₄BrNO₂S₂
MW: 242.11 g/mol
Melting Pt: 134…137 °C
MDL Number: MFCD00067990
CAS Number: 53595-65-6

Order Now

Specification Test Results

Melting point 134-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR