N-tert-Butyl 5-bromo-2-thiophenesulfonamide ≥97%

Supplier: Matrix Scientific

Synonyms: N-tert-Butyl 5-bromo-2-thiophenesulphonamide

004202-5G 004202-25G 004202-100G
101786-940EA 30.77 USD
101786-940 101786-942 101786-944
N-tert-Butyl 5-bromo-2-thiophenesulfonamide ≥97%
N-tert-Butyl 5-bromo-2-thiophenesulfonamide

Hazard: Irritant

Formula: C₈H₁₂BrNO₂S₂
MW: 298.22 g/mol
Melting Pt: 62 °C
Density: 1.527
MDL Number: MFCD01569259
CAS Number: 286932-39-6

Order Now

Specification Test Results

Melting point 62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR