4,4'-Diaminooctafluorobiphenyl ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2',3,3',5,5',6,6'-Octafluoro-[1,1'-biphenyl]-4,4'-diamine, Octafluorobenzidine
004185-1G
004185-5G
101786-908EA
100
USD
101786-908
101786-910
4,4'-Diaminooctafluorobiphenyl ≥97%
4,4'-Diaminooctafluorobiphenyl
TCA listed: T
Hazard: Irritant-Harmful
Formula:
C₁₂H₄F₈N₂ MW: 328.17 g/mol Melting Pt: 175…177 °C |
MDL Number:
MFCD00007646 CAS Number: 1038-66-0 |
Specification Test Results
Melting point | 175-177°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...