2,5-Dimethyl-4-methoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

004053-1G 004053-5G
101786-738EA 150.77 USD
101786-738 101786-740
2,5-Dimethyl-4-methoxyphenylacetonitrile ≥98%
2,5-Dimethyl-4-methoxyphenylacetonitrile

Hazard: Irritant

Formula: C₁₁H₁₃NO
Melting Pt: 54…56 °C
CAS Number: 105909-12-4

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR