2,3-Dimethyl-4-methoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

004052-1G 004052-5G
101786-734EA 76.92 USD
101786-734 101786-736
2,3-Dimethyl-4-methoxyphenylacetonitrile ≥98%
2,3-Dimethyl-4-methoxyphenylacetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₁H₁₃NO
Melting Pt: 77…80 °C
CAS Number: 206559-60-6

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR