3,5-Dimethoxy-4-hydroxyphenethylamine hydrochloride

Supplier: Matrix Scientific

003989-1G 003989-5G
101786-590EA 166.15 USD
101786-590 101786-592
3,5-Dimethoxy-4-hydroxyphenethylamine hydrochloride
3,5-Dimethoxy-4-hydroxyphenethylamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClNO₃
Melting Pt: 260…262 °C
MDL Number: MFCD03410251
CAS Number: 2176-14-9

Order Now

Specification Test Results

Melting point 260-262°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR