3,5-Dimethoxy-4-hydroxybenzhydrazide

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-3,5-dimethoxybenzhydrazide, 4-Hydroxy-3,5-dimethoxybenzohydrazide

003988-1G 003988-5G
101786-586EA 36.92 USD
101786-586 101786-588
3,5-Dimethoxy-4-hydroxybenzhydrazide
3,5-Dimethoxy-4-hydroxybenzhydrazide

Hazard: Irritant

Formula: C₉H₁₂N₂O₄
MW: 212.21 g/mol
Melting Pt: 156-157°C
MDL Number: MFCD00017077
CAS Number: 1443-76-1

Order Now

Specification Test Results

Melting point 156-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR