4,5-Dimethoxy-2-nitrotoluene ≥98%

Synonyms: 1,2-dimethoxy-4-methyl-5-nitrobenzene

003986-1G 003986-5G
101786-578 101786-580
4,5-Dimethoxy-2-nitrotoluene ≥98%
4,5-Dimethoxy-2-nitrotoluene

Hazard: Irritant

Formula: C₉H₁₁NO₄
MW: 197.187 g/mol
Melting Pt: 119…120 °C
MDL Number: MFCD00060708
CAS Number: 7509-11-7

Order Now

Specification Test Results

Melting point 119-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR