4,5-Dimethoxy-2-methylaniline
Supplier: Matrix Scientific
003985-1G
003985-5G
101786-574EA
36.92
USD
101786-574
101786-576
4,5-Dimethoxy-2-methylaniline
4,5-Dimethoxy-2-methylaniline
Hazard: Irritant/Irritant-Harmful
Formula:
C₉H₁₃NO₂ MW: 167.210 g/mol Melting Pt: 108…110 °C |
MDL Number:
MFCD00130067 CAS Number: 41864-45-3 |
Specification Test Results
Melting point | 108-110°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...