4,5-Dimethoxy-2-iodobenzoic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Iodo-4,5-dimethoxybenzoic acid
003984-1G
003984-5G
003984-25G
101786-570EA
84.62
USD
101786-570
101786-572
102550-836
4,5-Dimethoxy-2-iodobenzoic acid ≥98%
4,5-Dimethoxy-2-iodobenzoic acid
Hazard: Irritant
Formula:
C₉H₉IO₄ MW: 308.07 g/mol Melting Pt: 206…208 °C |
MDL Number:
MFCD00079751 CAS Number: 61203-48-3 |
Specification Test Results
Melting point | 206-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...